ContactUsLinkComponent
LCD Screen Holder ESD with Double Arm
LCD Screen Holder ESD with Double Arm
Catalog # SOVE93049002P
Supplier:  TRESTON
LCD Screen Holder ESD with Double Arm
Catalog # SOVE93049002P
Supplier:  TRESTON
Specifications
  • W×D:
    105×305 mm
  • Colour:
    Grey
  • Description:
    LCD screen holder with a double arm, suitable for VESA 75 and 100 standards.
  • Pk:
    1
Specifications
About this item
LCD screen holder with a double arm, suitable for VESA 75 and 100 standards.
  • Epoxy powder-coated steel frame
  • Arm length 200+200 mm, thickness of the arm 60 mm