ContactUsLinkComponent
Unibox Transport Boxes
Unibox Transport Boxes
Catalog # MULC45229UB160.12
Supplier:  MULTIROIR
Unibox Transport Boxes
Catalog # MULC45229UB160.12
Supplier:  MULTIROIR
Specifications
  • Description:
    Transport boxes, square
  • W×D×H:
    160×160×12,7 mm
  • Pk:
    90
Specifications
About this item
Unibox plastic box is used for transporting or packaging all types of products. Made of polypropylene, it is very resistant to shocks.
  • Closing, hinged, is done by pressure
Particularly suitable for industrial applications.