Orders LinkContactUsLinkComponent
Multi-Parameter Tester, CombiCheck 60
CombiCheck 60 Muliti-parameter standard solutions, 1 KIT
Catalog # 713-4101
Supplier:   WTW
Multi-Parameter Tester, CombiCheck 60
Catalog # 713-4101
Supplier:  WTW
CAS Number:  
Multi-Parameter Tester, CombiCheck 60
Multi-Parameter Tester, CombiCheck 60
CombiCheck 60 Muliti-parameter standard solutions, 1 KIT
CAS Number:  
Price
165,00 €
Log in for availability
1 * 1 KIT
Product Details & Documents
CombiCheck solutions are ready-to-use multi-parameter standards, Each package contains a standard solution and an addition solution. Both solutions can be used directly without dilution for quality assurance.
  • The standard solution is used to check the accuracy of the results for the complete system: procedure - analytical method - reagents - photometer
  • The addition solution is used to check sample-dependent influences by measuring the recovery rate and to determine the most suitable sample preparation method
The maximum number of determinations which can be made with a CombiCheck standard solution depends on the test set used. With the addition solution 280 determinations are always possible.
Parameters can be checked with their concentrations as follows:
COD: 250 mg/L COD
Chloride: 125 mg/L Cl
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    CombiCheck 60 Muliti-parameter standard solutions
Specifications