ContactUsLinkComponent
Swing Bin, Stainless Steel
Swing Bin, Stainless Steel
Catalog # TEKMHWBH-SB
Supplier:  TEKNOMEK
Swing Bin, Stainless Steel
Catalog # TEKMHWBH-SB
Supplier:  TEKNOMEK
Specifications
  • Capacity:
    90 L
  • Description:
    Swing bin with lid
  • W×D×H:
    490×340×810 mm
  • Pk:
    1
Specifications
About this item
Our waste bin with swing lid is commonly used in hygienic environments such as healthcare centres and laboratories for sanitary waste disposal.
  • 304L-grade stainless steel
  • Brushed dull polish finish
  • Removeable swing lid top
  • Fully enclosed
Featuring a swing lid design, our own manufactured waste bin comes fully welded with 304L-grade stainless steel construction. Its strength and secure build makes it ideal for depositing waste sharps.