Orders LinkContactUsLinkComponent
VWR® Filter Vials
VWR® Filter Vials
Supplier:  VWR Collection
Certificates
VWR® Filter Vials
10 Options Available Below
Product Details & Documents
Filter vials with membrane, PE snap ring cap (11 mm), with hole and silicone white/PTFE red septa.
Vials have different colour caps for different membranes: RC with grey caps, PVDF with yellow caps, Nylon with light-blue caps, PTFE with pink caps and PES with light-green caps.
Product Family Options
Product Information
  • Membrane
    Pore size
    Availability
    Price
  • Nylon
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • Nylon
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • PES (Polyethersulfone)
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • PES (Polyethersulfone)
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • PTFE (Polyetrafluoreoethylene)
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • PTFE (Polyetrafluoreoethylene)
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • PVDF (Polyvinylidene fluoride)
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • PVDF (Polyvinylidene fluoride)
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 247,00 €
    247,00 €
    1 * 100 items
       247,00 €
  • RC (Regenerated cellulose)
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 298,00 €
    298,00 €
    1 * 100 items
       298,00 €
  • RC (Regenerated cellulose)
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 100 items
    1 * 100 items - 298,00 €
    298,00 €
    1 * 100 items
       298,00 €