Pesticide reference standard, Dichlorophen 1000 µg/ml in methanol
Catalog # PROOU-EPA-1217
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:97-23-4
- Description:Dichlorophen 1,000 µg/ml in methanol
- Formula:C₁₃H₁₀Cl₂O₂
- Description:Dichlorophen