Anti-DERP2 Human Monoclonal Antibody [clone: H.AK6.A3.F4]
Catalog # NOVUDDX0210P-100
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:DERP2
- Clonality:Monoclonal
- Clone:H.AK6.A3.F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Human
- Isotype:IgG1 kappa
- Reactivity:Human
- Antigen synonyms:Allergen Der p II|Mite group 2 allergen Der p 2|Allergen=Der p 2|DPX|Allergen Der p2|DERP-2
- Storage buffer:Tris-NaCl (pH 8,0)
- Storage temperature:Store at −20 °C. Avoid freeze-thaw cycles.
- Concentration:0,5 mg/ml
- Immunogen:Derp-2 was used as immunogen.
- Purification:Ion exchange chromatography
- Pk:100 µG
Specifications
About this item
The DERP2 Antibody (H.AK6.A3.F4) from Novus Biologicals is a human monoclonal antibody to DERP2. This antibody reacts with human, other. The DERP2 Antibody (H.AK6.A3.F4) has been validated for the following applications: ELISA.
Type: Primary
Antigen: DERP2
Clonality: Monoclonal
Clone: H.AK6.A3.F4
Conjugation: Unconjugated
Epitope:
Host: Human
Isotype: IgG1 Kappa
Reactivity: Human, Other