Anti-MKK4 (Ser80) Phosphate Rabbit Polyclonal Antibody
CSTA9155S
- Antibody type:Primary
- Antigen symbol:MKK4 (Ser80) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Sequence:CERLRTHsIESSGK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: MKK4 (Ser80) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat