Anti-MOB1A (Thr12) Phosphate Rabbit Monoclonal Antibody [clone: D2E3]
CSTA8843S
- Antibody type:Primary
- Antigen symbol:MOB1A (Thr12) phosphate
- Clonality:Monoclonal
- Clone:D2E3
- Host:Rabbit
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Antigen synonyms:MOB1B (Thr12) phosphate
- Sequence:CSSRSSKtFKPKK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: MOB1A (Thr12) phosphate
Clonality: Monoclonal
Clone: D2E3
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Monkey