Anti-Acyl CoA synthase Rabbit Monoclonal Antibody [clone: D2H5]
Catalog # CSTA9189S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Acyl CoA synthase
- Clonality:Monoclonal
- Clone:D2H5
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CGVEVTSMKAMEDLGRANRRKPK
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Acyl CoA synthase
Clonality: Monoclonal
Clone: D2H5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey