Anti-RBPSUH Rabbit Monoclonal Antibody [clone: D10A4]
CSTA5313S
- Antibody type:Primary
- Antigen symbol:RBPSUH
- Clonality:Monoclonal
- Clone:D10A4
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- ChIP:Yes
- Sequence:CAFIGIGNSDQEMQQLNLEGKNY
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: RBPSUH
Clonality: Monoclonal
Clone: D10A4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey