Anti-APPL1 Rabbit Monoclonal Antibody [clone: D83H4]
Catalog # CSTA3858S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:APPL1
- Clonality:Monoclonal
- Clone:D83H4
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CGQSRPPTARTSSSGSLGSESTN
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: APPL1
Clonality: Monoclonal
Clone: D83H4
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey