Anti-Thymidylate Synthase Rabbit Polyclonal Antibody
Catalog # CSTA3766S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:Thymidylate Synthase
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Hamster,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CLKIQLQREPRPFPKLRILRKVEK
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: Thymidylate Synthase
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Hamster, Monkey