Anti-BiP Rabbit Monoclonal Antibody [clone: C50B12]
CSTA3177S
- Antibody type:Primary
- Antigen symbol:BiP
- Clonality:Monoclonal
- Clone:C50B12
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CKNQIGDKEKLGGKLSSEDKETMEKAVE
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: BiP
Clonality: Monoclonal
Clone: C50B12
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse