Anti-p14 Arf Mouse Monoclonal Antibody [clone: 4C6/4]
Catalog # CSTA2407S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p14 Arf
- Clonality:Monoclonal
- Clone:4C6/4
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: p14 Arf
Clonality: Monoclonal
Clone: 4C6/4
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human