Anti-XPB Mouse Monoclonal Antibody [clone: 2C6]
Catalog # CSTA8746S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:XPB
- Clonality:Monoclonal
- Clone:2C6
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:Recombinant fragment, corresponding to amino acids 500-782 of human ERCC3
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: XPB
Clonality: Monoclonal
Clone: 2C6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat, Monkey