Anti-GFAT2 Rabbit Monoclonal Antibody [clone: D40C7]
CSTA6917S
- Antibody type:Primary
- Antigen symbol:GFAT2
- Clonality:Monoclonal
- Clone:D40C7
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CATRRGSPLLIGVRSKYKLSTEQ
- Storage temperature:–20 °C
- Shipping temperature:Cold
- Pk:100 µl
Type: Primary
Antigen: GFAT2
Clonality: Monoclonal
Clone: D40C7
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human