Anti-THEX1 Rabbit Monoclonal Antibody [clone: D2B4]
CSTA4052S
- Antibody type:Primary
- Antigen symbol:THEX1
- Clonality:Monoclonal
- Clone:D2B4
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CSSLPIEGTPPPQMPHFRK
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: THEX1
Clonality: Monoclonal
Clone: D2B4
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human