Anti-TNF-R1 Rabbit Monoclonal Antibody [clone: C25C1]
Catalog # CSTA3736S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:TNF-R1
- Clonality:Monoclonal
- Clone:C25C1
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Seq:02
- Sequence:CSDPIPNPLQKWEDSAHKPQSLDTDDPA
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TNF-R1
Clonality: Monoclonal
Clone: C25C1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human