Anti-SHIP1 Rabbit Monoclonal Antibody [clone: C40G9]
Catalog # CSTA2727S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:SHIP1
- Clonality:Monoclonal
- Clone:C40G9
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Sequence:CTLSPDQQPTAWSYDQPPKDSPLGP
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SHIP1
Clonality: Monoclonal
Clone: C40G9
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human