Anti-eIF4G (Ser1147) phosphate Rabbit Polyclonal Antibody
Catalog # CSTA2441S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:eIF4G (Ser1147) phosphate
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Hamster,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CVVQRSSLsRERGE
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: eIF4G (Ser1147) phosphate
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat, Hamster, Monkey, Bovine