Anti-Lysine (K-Ac) acetylate Rabbit Monoclonal Antibody [clone: Ac-K²-100) MultiMab™ Rabbit mAb mix]
CSTA9814S
- Antibody type:Primary
- Antigen symbol:Lysine (K-Ac) acetylate
- Clonality:Monoclonal
- Clone:Ac−K2−100) MultiMab™ Rabbit mAb mix
- ELISA:Yes
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Multiple
- Western blot:Yes
- ChIP:Yes
- Sequence:CXXXX[K-Ac]XXX[K-Ac]XXXX[K-Ac]XXX[K-Ac]XXXX
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: Lysine (K-Ac) acetylate
Clonality: Monoclonal
Clone: Ac-K2-100) MultiMab™ Rabbit mAb mix
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: All