Anti-USP1 Rabbit Monoclonal Antibody [clone: D37B4]
Catalog # CSTA8033S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:USP1
- Clonality:Monoclonal
- Clone:D37B4
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CKVTEEKDFLNSLSPSTSPTSTPYL
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: USP1
Clonality: Monoclonal
Clone: D37B4
Conjugation:
Epitope:
Hoat: Rabbit
Isotype: IgG
Reactivity: Human,Mouse,Monkey