Anti-JMJD1B Mouse Monoclonal Antibody [clone: 6A1-1F5]
CSTA5377S
- Antibody type:Primary
- Antigen symbol:JMJD1B
- Clonality:Monoclonal
- Clone:6A1−1F5
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:Purified recombinant fragment of human JMJD1B (aa1467-aa1740) expressed in E. Coli.
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Type: Primary Antibodies
Antigen: JMJD1B
Clonality: Monoclonal
Clone: 6A1-1F5
Conjugation:
Epitope:
Hoat: Mouse
Isotype: IgG1
Reactivity: Human,Mouse,Rat,Monkey