Anti-p47phox Rabbit Monoclonal Antibody [clone: D21F6]
Catalog # CSTA4301S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:p47phox
- Clonality:Monoclonal
- Clone:D21F6
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Sequence:CASFLEPLDSPDETEDPEPNYAGE
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Size:100 µl
- Pk:100 µl
Specifications
About this item
Type: Primary Antibodies
Antigen: p47phox
Clonality: Monoclonal
Clone: D21F6
Conjugation:
Epitope:
Hoat: Rabbit
Isotype:
Reactivity: Human