Height Endurance Visor
Height endurance visor, Clear
Catalog # PRTWPA03CLR
Supplier: Portwest
Jump to:
Product Details & Documents
Panoramic wrap-around visor for Height Endurance helmets range. It allows the user to wear over prescription glasses.
- One-piece curved lens with moulded nose bridge
- Light and robust
- Anti-scratch coating for added durability
- Anti-fog coating for additional comfort to the wearer
- Retail box aids presentation for retail sales
Compatible with Height Endurance Helmets and suitable for wearing over most prescription glasses, this visor features an anti-scratch and anti-fog treatment for added durability.
Certifications: EN 170 (2C-1.2 ).
Specifications
- Colour:Clear
- Description:Height endurance visor