ContactUsLinkComponent
Combination Filter Bayonet Connection Systems, ABEK1P3
Combination Filter Bayonet Connection Systems, ABEK1P3
Catalog # PRTWP971BRR
Supplier:  Portwest
Combination Filter Bayonet Connection Systems, ABEK1P3
Catalog # PRTWP971BRR
Supplier:  Portwest
Specifications
  • Description:
    ABEK1P3 combination filter bayonet connection
  • Pk:
    4
Specifications
About this item
This combination gas and particle filter protects from organic and inorganic acid gases and vapours, sulphur dioxide, ammonia, and its derivatives.
  • Bayonet connection
  • Retail box aids presentation for retail sales
  • Lightweight and comfortable
Classified as a class 1 filter for gases and class 3 for particles, it protects against harmful substances, solvents, toxic particles, and micro-organisms. The special Portwest bayonet system ensures a safer connection between the mask and the filter. This filter is compatible with half masks P420, P430, P421, P431, and full face mask model P500.
Certifications: EN 14387 Type P Class 3R. CE-CAT III.