Anti-GAPDH Rabbit Monoclonal Antibody [clone: 14C10] (Biotinylated)
Catalog # CSTA5014S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:GAPDH
- Clonality:Monoclonal
- Clone:14C10
- Conjugation:Biotin
- Host:Rabbit
- Isotype:IgG
- Reactivity:Pig,Monkey,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Sequence:CRLEKPAKYDDIKKVVKQASEGPLKG
- Storage temperature:−20 °C
- Shipping temperature:Cold
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: GAPDH
Clonality: Monoclonal
Clone: 14C10
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey;Bovine;Pig