Anti-RagA Rabbit Monoclonal Antibody [clone: D8B5]
Catalog # CSTA4357S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:RagA
- Clonality:Monoclonal
- Clone:D8B5
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Sequence:CGGQDTFMENYFTSQRDN
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: RagA
Clonality: Monoclonal
Clone: D8B5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey