Anti-MyD88 Rabbit Monoclonal Antibody [clone: D80F5]
Catalog # CSTA4283S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:MyD88
- Clonality:Monoclonal
- Clone:D80F5
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster,Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CDFQTKFALSLSPGAHQKRLIPIKYKAMKK
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: MyD88
Clonality: Monoclonal
Clone: D80F5
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Hamster;Monkey