ContactUsLinkComponent
PU Foam Ear Plugs
PU Foam Ear Plugs
  PRTWEP02ORR
 :  Portwest
PU Foam Ear Plugs
  PRTWEP02ORR
 :  Portwest
  • Colour:
    Orange
  • Description:
    PU foam ear plugs
  • Pk:
    200 PAIR
PU foam disposable ear plugs made from soft and durable foam.
  • Disposable ear plugs made from soft PU foam
  • Supplied in pairs, within a handy portable dispenser box
  • One pair per polybag for hygienic storage
  • Easy to fit, with an excellent attenuation rate
  • Retail box aids presentation for retail sales
Easy to insert and remove. Excellent attenuation level and a comfortable fit for long periods of use. Each pair comes inside an individual hygienic polybag with fitting instructions. Individually packed for vending machines.
 : ANSI S3.19 (NRR 32dB). CE-CAT III.