Anti-Akt-phosphate Substrate (Ser/Thr) phosphate Rabbit Polyclonal Antibody
Supplier: Cell Signaling
Certificates
About this item
Type: Primary
Antigen: Akt-phosphate Substrate (Ser/Thr) phosphate
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Any species
Specifications
- Catalog No:
- CSTA9611S
- CSTA9611L
- Antigen symbol:
- Akt-phosphate Substrate (Ser/Thr) phosphate
- Akt-phosphate Substrate (Ser/Thr) phosphate
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Reactivity:
- Multiple
- Multiple
- Host:
- Rabbit
- Rabbit
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- Sequence:
- CXXXRXRXXtXXXX
- CXXXRXRXXtXXXX
- Storage temperature:
- −20 °C
- −20 °C
- Shipping temperature:
- Cold
- Cold
Specifications
Frequently Bought Together