Anti-Nestin Mouse Monoclonal Antibody
CSTA4760S
- Antibody type:Primary
- Antigen symbol:Nestin
- Clonality:Monoclonal
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Rat
- Seq:02
- Sequence:E15 rat spinal cord
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Nestin
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat