Anti-Tuberin/TSC2 Rabbit Polyclonal Antibody
CSTA3612S
- Antibody type:Primary
- Antigen symbol:Tuberin/TSC2
- Clonality:Polyclonal
- Host:Rabbit
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CQRKRLISSVEDFTEFV
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Type: Primary
Antigen: Tuberin/TSC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat;Monkey