Anti-SRC-1 Rabbit Monoclonal Antibody [clone: 128E7]
Catalog # CSTA2191S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:SRC-1
- Clonality:Monoclonal
- Clone:128E7
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- ChIP:Yes
- Sequence:CSPRRQVTSGLATRPRMPNNSFPPNISTLS
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: SRC-1
Clonality: Monoclonal
Clone: 128E7
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat;Monkey