Anti-TUG Rabbit Polyclonal Antibody
Catalog # CSTA2049S
Supplier: Cell Signaling
Specifications
- Antibody type:Primary
- Antigen symbol:TUG
- Clonality:Polyclonal
- Host:Rabbit
- ImmunoPrecipitation:Yes
- Reactivity:Monkey,Human,Rat,Mouse
- Western blot:Yes
- Sequence:CGTAQPVKRSLGKVPKWLKLPASKR
- Storage temperature:−20 °C
- Shipping temperature:Ambient
- Pk:100 µl
Specifications
About this item
Type: Primary
Antigen: TUG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat;Monkey