Order Entry
Slovenia
ContactUsLinkComponent
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane (Kryptofix® 221), Sigma-Aldrich®
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane (Kryptofix® 221), Sigma-Aldrich®
Catalog # SIAL291161-1G
Supplier:  Merck
CAS Number:  
4,7,13,16,21-Pentaoxa-1,10-diazabicyclo[8.8.5]tricosane (Kryptofix® 221), Sigma-Aldrich®
Catalog # SIAL291161-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :332.44 g/mol
Density:1.11 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ200
EINECS:250-592-1
CAS Number:31364-42-8
Formula:C₁₆H₃₂N₂O₅
MDL Number:MFCD00005108