Organic reference standard, Aniline 5000 µg/ml in methanol
Catalog # PROOU-EPA-1069
Supplier: LGC Standards PROMOCHEM
CAS Number:
Specifications
- Pk:1
- CAS No.:62-53-3
- Description:Aniline 5,000 µg/ml in methanol
- Formula:C₆H₇N
- Description:Aniline