ContactUsLinkComponent
Head, brain and arteries
Head, brain and arteries
Catalog # 763-0106
Supplier:  3B Scientific
undefined
Head, brain and arteries
Catalog # 763-0106
Supplier:  3B Scientific
Specifications
  • Description:
    Brain and arteries, in 8 parts, mounted on a head as base, C25
  • Pk:
    1
Specifications
About this item
The brain in a head base is delivered on a base. This C20 deluxe brain comes with opened head to allow detailed study of the brain's position in the skull. The human head is horizontally divided above the skull base. This medially divided deluxe brain model shows the brain arteries in detail and the basilar artery is removable. Both halves of this brain model can be disassembled.
  • Frontal with parietal lobes
  • Temporal with occipital lobes
  • Half of brain stem, half of cerebellum