Order Entry
Slovenia
ContactUsLinkComponent
Cy3 trans-cyclooctene fluorescent probe
Irritant
Cy3 trans-cyclooctene fluorescent probe
Catalog # AATB900
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Cy3 trans-cyclooctene fluorescent probe
Catalog # AATB900
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :940.26 g/mol
ADR:5,II
 :GHS07