ContactUsLinkComponent
Amplite® IR
Amplite® IR
Catalog # AATB11009
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® IR
Catalog # AATB11009
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    1 mg
  • Description:
    Amplite™ IR
Specifications
About this item
Our Amplite® IR is a fluorogenic peroxidase substrate that generates near infrared fluorescence upon reaction with peroxidase and H2O2.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.