ContactUsLinkComponent
Amplite® Fluorimetric Coenzyme A Quantitation Kit *Green Fluorescence*
Amplite® Fluorimetric Coenzyme A Quantitation Kit *Green Fluorescence*
Catalog # AATB15270
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Fluorimetric Coenzyme A Quantitation Kit *Green Fluorescence*
Catalog # AATB15270
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ fluorimetric coenzyme a quantitation kit *green fluorescence*
Specifications
About this item
Coenzyme A (CoA) is a universal and essential cofactor in all forms of cellular life acting as a principal acyl carrier in numerous biosynthetic, energy-yielding, and degradative pathways.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.