ContactUsLinkComponent
Amplite® Colorimetric Beta-Hydroxybutyrate (Ketone Body) Assay Kit
Amplite® Colorimetric Beta-Hydroxybutyrate (Ketone Body) Assay Kit
  AATB13830
 :  
undefined
Amplite® Colorimetric Beta-Hydroxybutyrate (Ketone Body) Assay Kit
  AATB13830
 :  
  • Pk:
    200 Tests
  • Description:
    Amplite™ colorimetric beta-hydroxybutyrate (ketone body) assay kit
Ketone bodies are produced by the liver and used peripherally as an energy source when blood glucose levels drop.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
 : For research use only.