ContactUsLinkComponent
Amplite® Human Serum Albumin (HSA) Site I Binding Assay Kit
Amplite® Human Serum Albumin (HSA) Site I Binding Assay Kit
Catalog # AATB25400
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Human Serum Albumin (HSA) Site I Binding Assay Kit
Catalog # AATB25400
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    200 Tests
  • Description:
    Amplite™ human serum albumin (HSA) site i binding assay kit
Specifications
About this item
Human serum albumin (HSA) is one of the most important carriers for acidic drugs in human plasma and has been shown to bind a large number of different compounds in a reversible manner.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.