ContactUsLinkComponent
Amplite® Colorimetric Tyrosinase Assay Kit
Amplite® Colorimetric Tyrosinase Assay Kit
Catalog # AATB11311
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Amplite® Colorimetric Tyrosinase Assay Kit
Catalog # AATB11311
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    Amplite™ colorimetric tyrosinase assay kit
Specifications
About this item
Tyrosinase is of great interest to drug discovery, life science research, food industry and cosmetics industry since it plays an important role in the biosynthetic pathway of melanin.
  • In-house QC testing
  • Sensitive
  • Suggested protocol available
Caution: For research use only.