Orders LinkContactUsLinkComponent
Screw Cap Tubes
Screw Cap Tubes
Supplier:   MEGRO
Screw Cap Tubes
Supplier:  MEGRO
Certificates
2 Options Available Below
Product Details & Documents
For secure transport and storage of samples.
  • Made of selected, biologically inert polypropylene
  • Reliable sealing
  • Polyethylene screw cap
  • Not autoclavable
Product Family Options
Product Information
  • Capacity
    Colour
    Length
    Tube ext.Ø
    Cap included
    Cap colour
    Availability
    Price
  • MEGR122120
    Screwcap tubes with enclosed cap
    Screwcap tubes with enclosed cap
    13 ml
    Natural
    100 mm
    16 mm
    Yes
    Natural
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 106,00 €
    106,00 €
    1 * 1.000 items
       106,00 €
  • MEGR122121
    Screwcap tubes with fitted cap
    Screwcap tubes with fitted cap
    13 ml
    Natural
    100 mm
    16 mm
    Yes
    Natural
    Log in for availability
    Log in for availability
    1 * 1.000 items
    1 * 1.000 items - 140,00 €
    140,00 €
    1 * 1.000 items
       140,00 €