ContactUsLinkComponent
Wall Mount for First Aid Kit, MT-CD
Wall Mount for First Aid Kit, MT-CD
Catalog # SOEH0301175
Supplier:  SOEHNGEN
Wall Mount for First Aid Kit, MT-CD
Catalog # SOEH0301175
Supplier:  SOEHNGEN
Specifications
  • Colour:
    Orange
  • Description:
    Wall bracket for first aid kit
  • Pk:
    1
Specifications
About this item
  • Material: ABS plastic
  • Dimensions: 400×300×150 mm
  • Shipping weight: 0,42 kg