ContactUsLinkComponent
StrandBrite™ Green Fluorimetric RNA Quantitation Kit
StrandBrite™ Green Fluorimetric RNA Quantitation Kit
Catalog # AATB17656
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
StrandBrite™ Green Fluorimetric RNA Quantitation Kit
Catalog # AATB17656
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    100 Tests
  • Description:
    StrandBrite™ green fluorimetric RNA quantitation kit
Specifications
About this item
Detecting and quantitating small amounts of RNA is extremely important for a wide variety of molecular biology procedures such as measuring yields of in vitro transcribed RNA and measuring RNA concentrations before performing Northern blot analysis, S1 nuclease assays, RNase protection assays, cDNA library preparation, reverse transcription PCR, and differential display PCR.
  • Highly sensitive RNA detection
  • Robust protocol
  • Multiple applications
Caution: For research use only.