ContactUsLinkComponent
Gelite™ Orange Nucleic Acid Gel Staining Kit
Gelite™ Orange Nucleic Acid Gel Staining Kit
Catalog # AATB17594
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Gelite™ Orange Nucleic Acid Gel Staining Kit
Catalog # AATB17594
Supplier:  AAT BIOQUEST
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    Gelite™ orange nucleic acid gel staining kit
Specifications
About this item
Gelite™ Orange is an extremely sensitive nucleic acid gel stain for detecting DNA or RNA in gels using a standard 300 nm UV transilluminator and Polaroid 667 black-and-white print film.
  • High sensitivity
  • Exceptional DNA affinity
  • Robust protocol
  • Multiple applications
Caution: For research use only.