Amplite® Colorimetric Caspase 3/7 Assay Kit *Yellow Color*
AATB13507
:
- Pk:200 Tests
- Description:Amplite™ colorimetric caspase 3/7 assay kit *yellow color*
- Tests per kit:200 Tests
Caspases play important roles in apoptosis and cell signaling.
- In-house QC testing
- Sensitive
- Suggested protocol available
: For research use only.